N-Methyl-N-(2-pyridin-2-ylethyl)amine ≥97%

Supplier: Matrix Scientific

Synonyms: 2-[2-(Methylamino)ethyl]pyridine

081894-1G 081894-10G
102836-064EA 76.92 USD
102836-064 102836-066
N-Methyl-N-(2-pyridin-2-ylethyl)amine ≥97%
N-Methyl-N-(2-pyridin-2-ylethyl)amine

Hazard: Corrosive/Air Sensitive

Formula: C₈H₁₂N₂
MW: 136.2 g/mol
Boiling Pt: 113…114 °C
Density: 0.984
Flash Pt: 97 °C
MDL Number: MFCD00006362
CAS Number: 5638-76-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR