N-Ethylpyridine-4-methylamine ≥97%
Supplier: Matrix Scientific
Synonyms:
N-(4-Pyridylmethyl)ethylamine, 4-(Ethylaminomethyl)pyridine
081891-1G
081891-10G
102836-056EA
44.62
USD
102836-056
102836-058
N-Ethylpyridine-4-methylamine ≥97%
N-Ethylpyridine-4-methylamine
Hazard: Irritant
Formula:
C₈H₁₂N₂ MW: 136.20 g/mol |
MDL Number:
MFCD00023632 CAS Number: 33403-97-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...