3-(Methylaminomethyl)pyridine ≥97%

Supplier: Matrix Scientific

081860-1G 081860-10G
102835-990EA 61.54 USD
102835-990 102835-992
3-(Methylaminomethyl)pyridine ≥97%
3-(Methylaminomethyl)pyridine

Hazard: Irritant-Harmful/Air Sensitive

Formula: C₇H₁₀N₂
Boiling Pt: 93…95 °C (12 mmHg)
MDL Number: MFCD00023610
CAS Number: 20173-04-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR