2-Bromobenzylamine hydrochloride ≥97%
Supplier: Matrix Scientific
081841-1G
081841-10G
102835-948EA
86.15
USD
102835-948
102835-950
2-Bromobenzylamine hydrochloride ≥97%
2-Bromobenzylamine hydrochloride
Hazard: Irritant
Formula:
C₇H₉BrClN MW: 222.52 g/mol |
MDL Number:
MFCD00012853 CAS Number: 5465-63-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...