tert-Butyl-3-(aminomethyl)benzylcarbamate ≥97%

Supplier: Matrix Scientific

Synonyms: tert-Butyl 3-(aminomethyl)benzylcarbamate

081827-1G 081827-10G
102835-918EA 192.31 USD
102835-918 102835-920
tert-Butyl-3-(aminomethyl)benzylcarbamate ≥97%
tert-Butyl-3-(aminomethyl)benzylcarbamate

Hazard: Irritant

Formula: C₁₃H₂₀N₂O₂
MW: 236.32 g/mol
MDL Number: MFCD01317800
CAS Number: 108467-99-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR