1-(N-Boc-aminomethyl)-4-(aminomethyl)benzene ≥97%
Supplier: Matrix Scientific
Synonyms:
tert-Butyl 4-(aminomethyl)benzylcarbamate
081826-1G
081826-10G
102835-914EA
184.62
USD
102835-914
102835-916
1-(N-Boc-aminomethyl)-4-(aminomethyl)benzene ≥97%
1-(N-Boc-aminomethyl)-4-(aminomethyl)benzene
Hazard: Irritant
Formula:
C₁₃H₂₀N₂O₂ MW: 236.3100 g/mol |
MDL Number:
MFCD02683058 CAS Number: 108468-00-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...