tert-Butyl-4-(hydroxymethyl)benzylcarbamate ≥97%

Supplier: Matrix Scientific

Synonyms: 4-(Aminomethyl)benzyl alcohol, N-BOC protected, 4-(Boc-aminomethyl-phenyl)-methanol, tert-Butyl 4-(hydroxymethyl)benzylcarbamate

081817-1G 081817-10G
102835-894EA 184.62 USD
102835-894 102835-896
tert-Butyl-4-(hydroxymethyl)benzylcarbamate ≥97%
tert-Butyl-4-(hydroxymethyl)benzylcarbamate

Hazard: Irritant

Formula: C₁₃H₁₉NO₃
MW: 237.3 g/mol
MDL Number: MFCD06411553
CAS Number: 123986-64-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR