(4-Dimethylaminobenzyl)methylamine ≥95%
Supplier: Matrix Scientific
Synonyms:
N,N-Dimethyl-4-[(methylamino)methyl]aniline
081809-1G
081809-10G
102835-868EA
276.92
USD
102835-868
102835-870
(4-Dimethylaminobenzyl)methylamine ≥95%
(4-Dimethylaminobenzyl)methylamine
Hazard: Irritant-Harmful
Formula:
C₁₀H₁₆N₂ MW: 164.25 g/mol |
MDL Number:
MFCD04600269 CAS Number: 83671-43-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...