3-(Methylamino)-1-phenylpropan-1-one hydrochloride ≥97%
Supplier: Matrix Scientific
081798-1G
081798-10G
102835-844EA
353.85
USD
102835-844
102835-846
3-(Methylamino)-1-phenylpropan-1-one hydrochloride ≥97%
3-(Methylamino)-1-phenylpropan-1-one hydrochloride
Hazard: Irritant
Formula:
C₁₀H₁₄ClNO Melting Pt: 139…141 °C |
MDL Number:
MFCD08063896 CAS Number: 2538-50-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...