DL-α-(Methylaminomethyl)benzyl alcohol ≥97%
Supplier: Matrix Scientific
081795-1G
081795-10G
102835-836EA
72.31
USD
102835-836
102835-838
DL-α-(Methylaminomethyl)benzyl alcohol ≥97%
DL-α-(Methylaminomethyl)benzyl alcohol
Hazard: Irritant
Formula:
C₉H₁₃NO MW: 151.21 g/mol |
MDL Number:
MFCD00004506 CAS Number: 6589-55-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...