3,4-Dimethoxybenzylamine ≥95%

Supplier: Matrix Scientific

081792-1G 081792-10G
102835-828EA 72.31 USD
102835-828 102835-830
3,4-Dimethoxybenzylamine ≥95%
3,4-Dimethoxybenzylamine

Hazard: Irritant

Formula: C₉H₁₃NO₂
MDL Number: MFCD00008116
CAS Number: 5763-61-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR