4-(Trifluoromethylthio)benzylamine ≥95%

Supplier: Matrix Scientific

081788-1G 081788-10G
102835-818EA 153.85 USD
102835-818 102835-820
4-(Trifluoromethylthio)benzylamine ≥95%
4-(Trifluoromethylthio)benzylamine

Hazard: Corrosive/Stored under Argon/Air Sensitive/Moisture Sensitive/Refrigerate

Formula: C₈H₈F₃NS
Boiling Pt: 224…225 °C
Density: 1.31
Flash Pt: 80 °C
MDL Number: MFCD00190129
CAS Number: 128273-56-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR