1-(2,4-Dimethoxyphenyl)methanamine hydrochloride ≥97%

Supplier: Matrix Scientific

Synonyms: (2,4-Dimethoxyphenyl)methanamine hydrochloride

081764-1G 081764-10G
102835-766EA 72.31 USD
102835-766 102835-768
1-(2,4-Dimethoxyphenyl)methanamine hydrochloride ≥97%
1-(2,4-Dimethoxyphenyl)methanamine hydrochloride

Hazard: Irritant

Formula: C₉H₁₄ClNO₂
MW: 203.67 g/mol
MDL Number: MFCD00012854
CAS Number: 20781-21-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR