(S)-(-)-β-Methylphenethylamine ≥97%

Supplier: Matrix Scientific

081762-1G 081762-10G
102835-760EA 138.46 USD
102835-760 102835-762
(S)-(-)-β-Methylphenethylamine ≥97%
(S)-(-)-β-Methylphenethylamine

Hazard: Irritant

Formula: C₉H₁₃N
MDL Number: MFCD00216741
CAS Number: 17596-79-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR