4-Methoxy-1-naphthaldehyde ≥97%
Supplier: Matrix Scientific
Synonyms:
4-Methoxynaphthalene-1-carbaldehyde
081748-1G
081748-10G
102835-722EA
72.31
USD
102835-722
102835-724
4-Methoxy-1-naphthaldehyde ≥97%
4-Methoxy-1-naphthaldehyde
Hazard: Irritant
Formula:
C₁₂H₁₀O₂ MW: 186.21 g/mol |
MDL Number:
MFCD00004006 CAS Number: 15971-29-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...