6-Chloro-1,3-dimethyl-1H-pyrazolo[3,4-b]pyridine5-carbaldehyde ≥97%

Supplier: Matrix Scientific

081742-1G 081742-10G
102835-706EA 430.77 USD
102835-706 102835-708
6-Chloro-1,3-dimethyl-1H-pyrazolo[3,4-b]pyridine5-carbaldehyde ≥97%
6-Chloro-1,3-dimethyl-1H-pyrazolo[3,4-b]pyridine5-carbaldehyde

Hazard: Irritant

Formula: C₉H₈ClN₃O
MW: 209.64 g/mol
Melting Pt: 170…173 °C
MDL Number: MFCD08691397
CAS Number: 898911-34-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR