(S)-Glyceraldehyde acetonide 50% in dichloromethane , 95%

Supplier: Matrix Scientific

081737-1G 081737-5G
102835-690EA 1040 USD
102835-690 102835-692
(S)-Glyceraldehyde acetonide 50% in dichloromethane , 95%
(S)-Glyceraldehyde acetonide

Hazard: Irritant/Refrigerate/Stored under Argon

Formula: C₆H₁₀O₃
MDL Number: MFCD00269683
CAS Number: 22323-80-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR