2-Butylimidazole-4-carbaldehyde ≥97%

Supplier: Matrix Scientific

Synonyms: 2-Butyl-1H-imidazole-5-carboxaldehyde

081706-1G 081706-10G
102835-606EA 127.69 USD
102835-606 102835-608
2-Butylimidazole-4-carbaldehyde ≥97%
2-Butylimidazole-4-carbaldehyde

Hazard: Irritant

Formula: C₈H₁₂N₂O
MW: 152.193 g/mol
MDL Number: MFCD01075030
CAS Number: 68282-49-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR