2-Ethyl-4-methyl-1H-imidazole-5-carbaldehyde ≥97%

Supplier: Matrix Scientific

081705-1G 081705-10G
102835-602EA 156.92 USD
102835-602 102835-604
2-Ethyl-4-methyl-1H-imidazole-5-carbaldehyde ≥97%
2-Ethyl-4-methyl-1H-imidazole-5-carbaldehyde

Hazard: Irritant

Formula: C₇H₁₀N₂O
MW: 138.17 g/mol
MDL Number: MFCD00173727
CAS Number: 88634-80-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR