4-Methyl-1H-imidazole-5-carbaldehyde ≥97%

Supplier: Matrix Scientific

Synonyms: 4-Methyl-5-imidazolecarboxaldehyde

081702-1G 081702-10G
102835-592EA 69.23 USD
102835-592 102835-594
4-Methyl-1H-imidazole-5-carbaldehyde ≥97%
4-Methyl-1H-imidazole-5-carbaldehyde

Hazard: Irritant

Formula: C₅H₆N₂O
MW: 110.12 g/mol
MDL Number: MFCD00173728
CAS Number: 68282-53-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR