1-(4-Iodophenyl)-1H-pyrrole-2-carbaldehyde ≥97%

Supplier: Matrix Scientific

081671-1G 081671-10G
102835-528EA 270.77 USD
102835-528 102835-530
1-(4-Iodophenyl)-1H-pyrrole-2-carbaldehyde ≥97%
1-(4-Iodophenyl)-1H-pyrrole-2-carbaldehyde

Hazard: Irritant

Formula: C₁₁H₈INO
MW: 297.1 g/mol
MDL Number: MFCD01764931
CAS Number: 261178-18-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR