5-[3,5-Di(trifluoromethyl)phenyl]-2-furaldehyde ≥97%

Supplier: Matrix Scientific

Synonyms: 5-[3,5-Bis(trifluoromethyl)phenyl]furan-2-carboxaldehyde

081664-1G 081664-10G
102835-506EA 127.69 USD
102835-506 102835-508
5-[3,5-Di(trifluoromethyl)phenyl]-2-furaldehyde ≥97%
5-[3,5-Di(trifluoromethyl)phenyl]-2-furaldehyde

Hazard: Irritant

Formula: C₁₃H₆F₆O₂
MW: 308.18 g/mol
MDL Number: MFCD00173833
CAS Number: 256658-04-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR