5-Nitro-2-furaldehyde ≥98%

Supplier: Matrix Scientific

Synonyms: 5-Nitrofurfural

081660-10G 081660-50G
102835-496EA 193.85 USD
102835-496 102835-498
5-Nitro-2-furaldehyde ≥98%
5-Nitro-2-furaldehyde

Hazard: Irritant

Formula: C₅H₃NO₄
MW: 141.08 g/mol
MDL Number: MFCD00003230
CAS Number: 698-63-5
UN: 1325
ADR: 4.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR