5-Chlorothiophene-2-carboxaldehyde ≥97%

Supplier: Matrix Scientific

Synonyms: 2-Chloro-5-thiophenecarboxaldehyde, 2-Chloro-5-formylthiophene

081658-1G 081658-10G
102835-490EA 101.54 USD
102835-490 102835-492
5-Chlorothiophene-2-carboxaldehyde ≥97%
5-Chlorothiophene-2-carboxaldehyde

Hazard: Irritant

Formula: C₅H₃ClOS
MW: 146.59 g/mol
MDL Number: MFCD00047090
CAS Number: 7283-96-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR