2,6-Pyridinedicarboxaldehyde ≥97%

Supplier: Matrix Scientific

081625-1G 081625-10G
102835-416EA 566.15 USD
102835-416 102835-418
2,6-Pyridinedicarboxaldehyde ≥97%
2,6-Pyridinedicarboxaldehyde

Hazard: Irritant

Formula: C₇H₅NO₂
MW: 135.12 g/mol
MDL Number: MFCD00010103
CAS Number: 5431-44-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR