4-(Benzyloxy)-3-chlorobenzaldehyde ≥97%

Supplier: Matrix Scientific

081600-1G 081600-10G
102835-368EA 143.08 USD
102835-368 102835-370
4-(Benzyloxy)-3-chlorobenzaldehyde ≥97%
4-(Benzyloxy)-3-chlorobenzaldehyde

Hazard: Irritant

Formula: C₁₄H₁₁ClO₂
MDL Number: MFCD00831645
CAS Number: 66422-84-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR