4-Methoxy-3-nitrobenzaldehyde ≥97%

Supplier: Matrix Scientific

081595-1G 081595-10G 081595-50G
102835-354EA 56.92 USD
102835-354 102835-356 102835-358
4-Methoxy-3-nitrobenzaldehyde ≥97%
4-Methoxy-3-nitrobenzaldehyde

Hazard: Irritant

Formula: C₈H₇NO₄
MW: 181.15 g/mol
MDL Number: MFCD00126498
CAS Number: 31680-08-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR