2,6-Dimethoxybenzaldehyde ≥97%

Supplier: Matrix Scientific

081589-10G 081589-50G
102835-336EA 140 USD
102835-336 102835-338
2,6-Dimethoxybenzaldehyde ≥97%
2,6-Dimethoxybenzaldehyde

Hazard: Irritant

Formula: C₉H₁₀O₃
MW: 166.18 g/mol
Boiling Pt: 285 °C
Melting Pt: 96…98 °C
MDL Number: MFCD00010862
CAS Number: 3392-97-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR