2,4-Dimethoxy-6-methylbenzaldehyde ≥97%

Supplier: Matrix Scientific

081588-1G 081588-10G
102835-332EA 283.08 USD
102835-332 102835-334
2,4-Dimethoxy-6-methylbenzaldehyde ≥97%
2,4-Dimethoxy-6-methylbenzaldehyde

Hazard: Irritant

Formula: C₁₀H₁₂O₃
MDL Number: MFCD00033851
CAS Number: 7149-90-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR