4-[(4-Methylphenyl)thio]-3-nitrobenzaldehyde ≥95%

Supplier: Matrix Scientific

081575-1G 081575-10G
102835-302EA 101.54 USD
102835-302 102835-304
4-[(4-Methylphenyl)thio]-3-nitrobenzaldehyde ≥95%
4-[(4-Methylphenyl)thio]-3-nitrobenzaldehyde

Hazard: Irritant

Formula: C₁₄H₁₁NO₃S
MDL Number: MFCD00098766
CAS Number: 270262-89-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR