4,6-Dimethoxysalicylaldehyde ≥97%

Supplier: Matrix Scientific

081561-1G 081561-10G
102835-272EA 353.85 USD
102835-272 102835-274
4,6-Dimethoxysalicylaldehyde ≥97%
4,6-Dimethoxysalicylaldehyde

Hazard: Irritant

Formula: C₉H₁₀O₄
MDL Number: MFCD00003328
CAS Number: 708-76-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR