2-Fluoro-4-methylbenzaldehyde ≥97%
Supplier: Matrix Scientific
Synonyms:
2-Fluoro-p-tolylaldehyde, 2-Fluoro-4-tolualdehyde, 2-Fluoro-p-tolualdehyde
081560-1G
081560-10G
102835-268EA
49.23
USD
102835-268
102835-270
2-Fluoro-4-methylbenzaldehyde ≥97%
2-Fluoro-4-methylbenzaldehyde
Hazard: Irritant
Formula:
C₈H₇FO |
MDL Number:
MFCD03094325 CAS Number: 146137-80-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...