2-Fluoro-m-anisaldehyde ≥97%

Supplier: Matrix Scientific

081558-1G 081558-10G
102835-262EA 127.69 USD
102835-262 102835-264
2-Fluoro-m-anisaldehyde ≥97%
2-Fluoro-m-anisaldehyde

Hazard: Irritant

Formula: C₈H₇FO₂
MDL Number: MFCD03411962
CAS Number: 103438-88-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR