2,5-Bis(trifluoromethyl)benzaldehyde ≥97%

Supplier: Matrix Scientific

081557-1G 081557-10G
102835-258EA 15.38 USD
102835-258 102835-260
2,5-Bis(trifluoromethyl)benzaldehyde ≥97%
2,5-Bis(trifluoromethyl)benzaldehyde

Hazard: Irritant

Formula: C₉H₄F₆O
MW: 242.12 g/mol
MDL Number: MFCD00674087
CAS Number: 395-64-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR