4-(3-Bromopropoxy)benzaldehyde ≥97%

Supplier: Matrix Scientific

081555-1G 081555-10G
102835-252EA 353.85 USD
102835-252 102835-254
4-(3-Bromopropoxy)benzaldehyde ≥97%
4-(3-Bromopropoxy)benzaldehyde

Hazard: Irritant

Formula: C₁₀H₁₁BrO₂
MDL Number: MFCD08447768
CAS Number: 17954-81-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR