5-Hydroxy-3,4-dimethoxybenzaldehyde ≥97%
Supplier: Matrix Scientific
Synonyms:
3-Hydroxy-4,5-dimethoxybenzaldehyde, 3,4-Dimethoxy-5-hydroxybenzaldehyde
081536-1G
081536-10G
102835-206EA
169.23
USD
102835-206
102835-208
5-Hydroxy-3,4-dimethoxybenzaldehyde ≥97%
5-Hydroxy-3,4-dimethoxybenzaldehyde
Hazard: Irritant
Formula:
C₉H₁₀O₄ MW: 182.1733 g/mol Melting Pt: 67-69°C |
MDL Number:
MFCD00010442 CAS Number: 29865-90-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...