2-Methoxy-5-(trifluoromethyl)benzaldehyde ≥97%

Supplier: Matrix Scientific

081533-1G 081533-10G
102835-198EA 21.54 USD
102835-198 102835-200
2-Methoxy-5-(trifluoromethyl)benzaldehyde ≥97%
2-Methoxy-5-(trifluoromethyl)benzaldehyde

Hazard: Irritant

Formula: C₉H₇F₃O₂
MW: 204.15 g/mol
MDL Number: MFCD03788528
CAS Number: 146539-83-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR