3-(Oxiran-2-ylmethoxy)benzaldehyde ≥97%

Supplier: Matrix Scientific

081531-1G 081531-10G
102835-192EA 115.38 USD
102835-192 102835-194
3-(Oxiran-2-ylmethoxy)benzaldehyde ≥97%
3-(Oxiran-2-ylmethoxy)benzaldehyde

Hazard: Irritant

Formula: C₁₀H₁₀O₃
MDL Number: MFCD00203974
CAS Number: 22590-64-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR