2-[4-(tert-Butyl)phenoxy]-5-nitrobenzaldehyde ≥97%

Supplier: Matrix Scientific

081530-1G 081530-10G
102835-188EA 241.54 USD
102835-188 102835-190
2-[4-(tert-Butyl)phenoxy]-5-nitrobenzaldehyde ≥97%
2-[4-(tert-Butyl)phenoxy]-5-nitrobenzaldehyde

Hazard: Irritant

Formula: C₁₇H₁₇NO₄
MDL Number: MFCD00084918
CAS Number: 175278-47-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR