2,5-Dihydroxybenzaldehyde ≥98%

Supplier: Matrix Scientific

081524-1G 081524-10G 081524-50G
102835-170EA 44.62 USD
102835-170 102835-172 102835-174
2,5-Dihydroxybenzaldehyde ≥98%
2,5-Dihydroxybenzaldehyde

Hazard: Irritant/Air Sensitive/Stored under Argon

Formula: C₇H₆O₃
MW: 138.12 g/mol
Melting Pt: 97-103°C
MDL Number: MFCD00003333
CAS Number: 1194-98-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR