2-Amino-1-methoxypropane ≥97%

Supplier: Matrix Scientific

081520-1G 081520-10G
102835-156EA 120 USD
102835-156 102835-158
2-Amino-1-methoxypropane ≥97%
2-Amino-1-methoxypropane

Hazard: Flammable/Corrosive

Formula: C₄H₁₁NO
MW: 89.14 g/mol
Boiling Pt: 92-93°C
Density: 0.845
Flash Pt: 9°C
MDL Number: MFCD00008084
CAS Number: 37143-54-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR