Bis(aminoguanidinium) sulfate ≥97%
Supplier: Matrix Scientific
081518-1G
081518-10G
102835-150EA
44.62
USD
102835-150
102835-152
Bis(aminoguanidinium) sulfate ≥97%
Bis(aminoguanidinium)sulphate
Hazard: Irritant
Formula:
C₂H₁₄N₈O₄S Melting Pt: 207°C |
MDL Number:
MFCD00035665 CAS Number: 996-19-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...