4-Ethylamino-1-butanol ≥97%

Supplier: Matrix Scientific

081515-1G 081515-10G
102835-136EA 156.92 USD
102835-136 102835-138
4-Ethylamino-1-butanol ≥97%
4-Ethylamino-1-butanol

Hazard: Irritant

Formula: C₆H₁₅NO
MDL Number: MFCD00671534
CAS Number: 39216-86-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR