N-tert-Butylethylamine ≥97%

Supplier: Matrix Scientific

081513-1G 081513-10G
102835-128EA 101.54 USD
102835-128 102835-130
N-tert-Butylethylamine ≥97%
N-tert-Butylethylamine

Hazard: Flammable/Corrosive

Formula: C₆H₁₅N
Boiling Pt: 89 °C
Density: 0.73
MDL Number: MFCD00059367
CAS Number: 4432-77-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR