1-Amino-2-methylbutane ≥97%

Supplier: Matrix Scientific

Synonyms: 2-Methylbutylamine, 2-Methyl-1-butylamine

081512-1G 081512-10G
102835-124EA 101.54 USD
102835-124 102835-126
1-Amino-2-methylbutane ≥97%
1-Amino-2-methylbutane

Hazard: Irritant

Formula: C₅H₁₃N
MW: 87.16 g/mol
MDL Number: MFCD00008147
CAS Number: 96-15-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR