N-Benzyloxycarbonyl-D-alaninol ≥97%

Supplier: Matrix Scientific

Synonyms: (2R)-N-Benzyloxycarbonylamino-1-propanol, N-Cbz-D-Alaninol, Z-D-Alaninol, (R)-2-(Cbz-amino)-1-propanol

081510-1G 081510-10G
102835-118EA 127.69 USD
102835-118 102835-120
N-Benzyloxycarbonyl-D-alaninol ≥97%
N-Cbz-D-Alaninol

Hazard: Irritant

Formula: C₁₁H₁₅NO₃
MW: 209.2 g/mol
MDL Number: MFCD00672531
CAS Number: 61425-27-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR