N-(2-Aminoethyl)-1,3-propanediamine ≥97%
Supplier: Matrix Scientific
Synonyms:
3-(2-Aminoethylamino)propylamine
081509-1G
081509-10G
102835-114EA
44.62
USD
102835-114
102835-116
N-(2-Aminoethyl)-1,3-propanediamine ≥97%
N-(2-Aminoethyl)-1,3-propanediamine
Hazard: Irritant
Formula:
C₅H₁₅N₃ |
MDL Number:
MFCD00008211 CAS Number: 13531-52-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...