8-Amino-2-naphthol ≥97%
Supplier: Matrix Scientific
Synonyms:
1-Amino-7-naphthol
081501-1G
081501-10G
102835-090EA
15.38
USD
102835-090
102835-092
8-Amino-2-naphthol ≥97%
8-Amino-2-naphthol
Hazard: Irritant
Formula:
C₁₀H₉NO MW: 159.19 g/mol |
MDL Number:
MFCD00004031 CAS Number: 118-46-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...