N-Methyl-N-1-naphthylamine ≥97%

Supplier: Matrix Scientific

Synonyms: N-Methylnaphthalen-1-amine

081490-1G 081490-10G
102835-060EA 241.54 USD
102835-060 102835-062
N-Methyl-N-1-naphthylamine ≥97%
N-Methyl-N-1-naphthylamine

Hazard: Irritant

Formula: C₁₁H₁₁N
MW: 157.22 g/mol
MDL Number: MFCD00972770
CAS Number: 2216-68-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR