3-Amino-5-bromo-1-methyl-1H-indazole ≥97%

Supplier: Matrix Scientific

081484-1G 081484-10G
102835-044EA 636.92 USD
102835-044 102835-046
3-Amino-5-bromo-1-methyl-1H-indazole ≥97%
3-Amino-5-bromo-1-methyl-1H-indazole

Hazard: Irritant

Formula: C₈H₈BrN₃
MW: 226.08 g/mol
MDL Number: MFCD09864804
CAS Number: 1000018-06-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR